Behenic acid ≥99.0%
Supplier: TCI America
Synonyms:
Docosanoic acid
B1747-5G
TCB1747-5GEA
174.58
USD
TCB1747-5G
Behenic acid ≥99.0%
Behenic acid
CAS Number: 112-85-6
MDL Number: MFCD00002807
Molecular Formula: C22H44O2
Molecular Weight: 340.59
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 306
Melting point (°C): 81
Flash Point (°C): 228
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Docosanoic Acid
Formula:
C₂₂H₄₄O₂ MW: 340.59 g/mol Boiling Pt: 306 °C Melting Pt: 81 °C Flash Pt: 228 °C |
MDL Number:
MFCD00002807 CAS Number: 112-85-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...