1-tert-Butyl-3-ethylbenzene ≥98.0%

Supplier: TCI America

B1739-5ML
TCB1739-005MLEA 54 USD
TCB1739-005ML
1-tert-Butyl-3-ethylbenzene ≥98.0%
1-tert-Butyl-3-ethylbenzene

CAS Number: 14411-56-4
MDL Number: MFCD00060845
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 38
Specific Gravity (20/20): 0.86

Formula: C₁₂H₁₈
Boiling Pt: 205 °C
Density: 0,86 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00060845
CAS Number: 14411-56-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR