Bis[4-(3-aminophenoxy)phenyl] Sulfone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B1681-25G
TCB1681-025GEA 24 USD InStock
TCB1681-025G
Bis[4-(3-aminophenoxy)phenyl] Sulfone ≥98.0% (by HPLC, titration analysis)
Bis[4-(3-aminophenoxy)phenyl] sulfone

CAS Number: 30203-11-3
MDL Number: MFCD00054738
Molecular Formula: C24H20N2O4S
Molecular Weight: 432.49
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 135

Formula: C₂₄H₂₀N₂O₄S
MW: 432.49 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00054738
CAS Number: 30203-11-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR