N-Boc-L-threonine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Boc-Thr-OH
B1638-10G
TCB1638-010GEA
518
USD
TCB1638-010G
N-Boc-L-threonine ≥98.0% (by titrimetric analysis)
N-Boc-L-threonine
CAS Number: 2592-18-9
MDL Number: MFCD00065946
Molecular Formula: C9H17NO5
Molecular Weight: 219.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 84
Specific rotation [a]20/D: -7 deg (C=1, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-threonine, Boc-Thr-OH
Formula:
C9H17NO5 MW: 219.2 g/mol Melting Pt: 84 °C |
MDL Number:
MFCD00065946 CAS Number: 2592-18-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...