4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Benzyl N-Boc-L-aspartateN-Boc-L-aspartic Acid 4-Benzyl EsterN-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl EsterBoc-Asp(OBzl)-OH

B1628-25G B1628-5G
TCB1628-025GEA 147.81 USD
TCB1628-025G TCB1628-005G
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by titrimetric analysis)
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

CAS Number: 7536-58-5
MDL Number: MFCD00065564
Molecular Formula: C16H21NO6
Molecular Weight: 323.35
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 103
Specific rotation [a]20/D: -20 deg (C=2, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Benzyl N-Boc-L-aspartate, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester, Boc-Asp(OBzl)-OH

Formula: C₁₆H₂₁NO₆
MW: 323.35 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00065564
CAS Number: 7536-58-5
https://us.vwr.com/stibo/web/std.lang.all/33/89/28933389.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR