N-ɑ-Boc-L-asparagine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Boc-Asn-OHBoc-L-Asparagine

B1627-10G
TCB1627-010GEA 25 USD
TCB1627-010G
N-ɑ-Boc-L-asparagine ≥98.0% (by HPLC, titration analysis)
N-alpha-Boc-L-asparagine

CAS Number: 7536-55-2
MDL Number: MFCD00038152
Molecular Formula: C9H16N2O5
Molecular Weight: 232.24
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 166
Specific rotation [a]20/D: -7 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-L-asparagine, Boc-Asn-OH

Formula: C9H16N2O5
MW: 232.24 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00038152
CAS Number: 7536-55-2
https://us.vwr.com/stibo/web/std.lang.all/33/88/28933388.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR