4'-Bromo-2'-methylacetanilide ≥98.0%

Supplier: TCI America

B1556-25G
TCB1556-025GEA 78 USD
TCB1556-025G
4'-Bromo-2'-methylacetanilide ≥98.0%
4'-Bromo-2'-methylacetanilide

CAS Number: 24106-05-6
MDL Number: MFCD00017779
Molecular Formula: C9H10BrNO
Molecular Weight: 228.09
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 159

https://us.vwr.com/stibo/web/std.lang.all/33/24/28933324.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR