Methyl-3-benzoylpropionate ≥98.0%

Supplier: TCI America

Synonyms: 4-Oxo-4-phenylbutyric Acid Methyl Ester3-Benzoylpropionic Acid Methyl EsterMethyl 4-Oxo-4-phenylbutyrateMethyl 4-oxo-4-phenylbutanoate

B1546-5G B1546-25G
TCB1546-005GEA 76.13 USD
TCB1546-005G TCB1546-025G
Methyl-3-benzoylpropionate ≥98.0%
Methyl-3-benzoylpropionate

CAS Number: 25333-24-8
MDL Number: MFCD00014906
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: 3-Benzoylpropionic Acid Methyl Ester, Methyl 4-Oxo-4-phenylbutyrate, 4-Oxo-4-phenylbutyric Acid Methyl Ester

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Density: 1.14 (20 °C)
MDL Number: MFCD00014906
CAS Number: 25333-24-8
https://us.vwr.com/stibo/web/std.lang.all/33/16/28933316.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR