(S)-1-Benzyl-3-pyrrolidinol ≥98.0%
Synonyms:
(S)-1-Benzyl-3-hydroxypyrrolidine
CAS Number: 101385-90-4
MDL Number: MFCD00075485
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: 4.6 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-1-Benzyl-3-hydroxypyrrolidine
Formula:
C₁₁H₁₅NO MW: 177.25 g/mol Boiling Pt: 120 °C Density: 1.07 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00075485 CAS Number: 101385-90-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...