1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene hydrate ≥98.0%
B1419-25G
B1419-5G
TCB1419-005GEA
103
USD
TCB1419-005G
TCB1419-025G
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene hydrate ≥98.0%
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene hydrate
CAS Number: 1992-15-0
MDL Number: MFCD00192487
Molecular Formula: C12H6F12O2
Molecular Weight: 410.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 94
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Bis(alpha-hydroxyhexafluoroisopropyl)benzene, alpha,alpha,alpha',alpha'-Tetrakis(trifluoromethyl)-1,4-benzenedimethanol
Formula:
C₁₂H₆F₁₂O₂·H₂O Melting Pt: 94 °C |
MDL Number:
MFCD00192487 CAS Number: 1992-15-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...