Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-DIOP ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (R,R)-DIOP(4R,5R)-(-)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane

B1113-1G
TCB1113-001GEA 136.29 USD InStock
TCB1113-001G
(-)-DIOP ≥97.0% (by titrimetric analysis)
(-)-DIOP
CAS Number: 32305-98-9
MDL Number: MFCD00014107
Molecular Formula: C31H32O2P2
Molecular Weight: 498.54
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 90
Specific rotation [a]20/D: -6 deg (C=2, Toluene)

Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (4R,5R)-(-)-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (-)-DIOP, (2R,3R)-(-)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane


Formula: C₃₁H₃₂O₂P₂
MW: 498,532 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00014107
CAS Number: 32305-98-9
EINECS: 250-984-2
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty