Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

(+)-DIOP ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (4S,5S)-(+)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane(S,S)-DIOP

B1112-1G
TCB1112-001GEA 126.93 USD InStock
TCB1112-001G
(+)-DIOP ≥95.0% (by GC, titration analysis)
(+)-DIOP
CAS Number: 37002-48-5
MDL Number: MFCD00009760
Molecular Formula: C31H32O2P2
Molecular Weight: 498.54
Purity/Analysis Method: 95.0% (GC,T)
Form: Crystal
Melting point (°C): 90

Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (4S,5S)-(+)-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (+)-DIOP, (2S,3S)-(+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane


Formula: C₃₁H₃₂O₂P₂
MW: 498,532 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00009760
CAS Number: 37002-48-5
EINECS: 253-307-9
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty