2,5-Bis(4-(diethylamino)phenyl)-1,3,4-oxadiazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,5-Bis(diethylamino)phenyl-1,3,4-oxadiazole
B1066-25G
TCB1066-025GEA
66
USD
TCB1066-025G
2,5-Bis(4-(diethylamino)phenyl)-1,3,4-oxadiazole ≥98.0% (by titrimetric analysis)
2,5-Bis(4-(diethylamino)phenyl)-1,3,4-oxadiazole
CAS Number: 1679-98-7
MDL Number: MFCD00010070
Molecular Formula: C22H28N4O
Molecular Weight: 364.49
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 150
Formula:
C₂₂H₂₈N₄O Melting Pt: 150 °C |
MDL Number:
MFCD00798595 CAS Number: 1679-98-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...