1,4-Bis(3-aminopropyl)piperazine ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Di(3-aminopropyl)piperazine

B1041-500ML B1041-25ML
TCB1041-500MLEA 232 USD
TCB1041-500ML TCB1041-25ML
1,4-Bis(3-aminopropyl)piperazine ≥98.0%
1,4-Bis(3-aminopropyl)piperazine

CAS Number: 7209-38-3
MDL Number: MFCD00006161
Molecular Formula: C10H24N4
Molecular Weight: 200.33
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 168
Freezing point (°C): 14
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 1,4-Di(3-aminopropyl)piperazine

Formula: C₁₀H₂₄N₄
MW: 200.33 g/mol
Boiling Pt: 158 °C
Density: 0.97 (20 °C)
Flash Pt: 168 °C
MDL Number: MFCD00006161
CAS Number: 7209-38-3
https://us.vwr.com/stibo/web/std.lang.all/29/04/28932904.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR