N-Benzylideneaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1034-25G
TCB1034-025GEA 71.58 USD
TCB1034-025G
N-Benzylideneaniline ≥98.0% (by GC, titration analysis)
N-Benzylideneaniline

CAS Number: 538-51-2
MDL Number: MFCD00003027
Molecular Formula: C13H11N
Molecular Weight: 181.24
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 52
Flash Point (°C): 110


Other Product Names / Synonyms: Benzalaniline

Formula: C₁₃H₁₁N
MW: 181,24
Boiling Pt: 300 °C
Melting Pt: 52 °C
Flash Pt: 110 °C
MDL Number: MFCD00003027
CAS Number: 538-51-2
EINECS: 208-694-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR