Boc-ON ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide
CAS Number: 58632-95-4
MDL Number: MFCD00001863
Molecular Formula: C13H14N2O3
Molecular Weight: 246.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 88
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: 2-Boc-oxyimino-2-phenylacetonitrile
Formula:
C13H14N2O3 MW: 246.27 g/mol Melting Pt: 88 °C |
MDL Number:
MFCD00001863 CAS Number: 58632-95-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...