Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-tert-Butoxycarbonylimidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: N-Boc-imidazole

B0916-10G
TCB0916-010G
N-tert-Butoxycarbonylimidazole ≥98.0% (by titrimetric analysis)
CAS Number: 49761-82-2
MDL Number: MFCD00014497
Molecular Formula: C8H12N2O2
Molecular Weight: 168.20
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 64
Melting point (°C): 46

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: N-Boc-imidazole


Formula: C₈H₁₂N₂O₂
MW: 168.20 g/mol
Boiling Pt: 64 °C
Melting Pt: 46 °C
MDL Number: MFCD00014497
CAS Number: 49761-82-2
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)
Boiling point (°C) 64
Melting point (°C) 46


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty