N-Benzoylthiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-CarbamothioylbenzamideN-(Carbamothioyl)benzamide

B0802-25G
TCB0802-025GEA 93.26 USD
TCB0802-025G
N-Benzoylthiourea ≥98.0% (by HPLC, total nitrogen)
N-Benzoylthiourea

CAS Number: 614-23-3
MDL Number: MFCD00041191
Molecular Formula: C8H8N2OS
Molecular Weight: 180.23
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 173

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00041191
CAS Number: 614-23-3
https://us.vwr.com/stibo/web/std.lang.all/27/13/28932713.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR