Butylmagnesium chloride 23% ∼2 mol/L in THF

Synonyms: n-Butylmagnesium chloride

B0726-250G
TCB0726-250GEA 76.59 USD
TCB0726-250G
Butylmagnesium chloride 23% ∼2 mol/L in THF
Butylmagnesium chloride

CAS Number: 693-04-9
MDL Number: MFCD00000475
Molecular Formula: C4H9ClMg
Molecular Weight: 116.87
Flash Point (°C): -14
Specific Gravity (20/20): 0.97

Formula: C₄H₉ClMg
MW: 116.87 g/mol
Density: 0,97 (20 °C)
Flash Pt: –14 °C
MDL Number: MFCD00000475
CAS Number: 693-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR