4-Bromophenyl isocyanate ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromo-4-isocyanatobenzene

B0636-25G B0636-5G
TCB0636-25GEA 457 USD
TCB0636-25G TCB0636-5G
4-Bromophenyl isocyanate ≥98.0%
4-Bromophenyl isocyanate

CAS Number: 2493-02-9
MDL Number: MFCD00002022
Molecular Formula: C7H4BrNO
Molecular Weight: 198.02
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 41
Flash Point (°C): 109
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid 4-Bromophenyl Ester

Formula: C₇H₄BrNO
MW: 198.02 g/mol
Boiling Pt: 158 °C
Melting Pt: 41 °C
Flash Pt: 109 °C
MDL Number: MFCD00002022
CAS Number: 2493-02-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR