2-Bromohexanoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
DL-2-Bromohexanoic acid
B0601-500G
B0601-25G
TCB0601-25GEA
23
USD
TCB0601-25G
TCB0601-500G
2-Bromohexanoic acid ≥98.0% (by GC, titration analysis)
2-Bromohexanoic acid
CAS Number: 616-05-7
MDL Number: MFCD00004218
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 110
Specific Gravity (20/20): 1.39
Formula:
C₆H₁₁BrO₂ MW: 195.06 g/mol Boiling Pt: 240 °C Density: 1.39 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004218 CAS Number: 616-05-7 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...