L(-)-Bornyl acetate ≥70.0% (contains ca. 20% Isobornyl Acetate)

Supplier: TCI America

B0526-25ML
TCB0526-025MLEA 20.1 USD
TCB0526-025ML
L(-)-Bornyl acetate ≥70.0% (contains ca. 20% Isobornyl Acetate)
L(-)-Bornyl acetate

CAS Number: 5655-61-8
MDL Number: MFCD00135944
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: 70.0% (GC)
Form: Clear Liquid
Flash Point (°C): 84
Specific Gravity (20/20): 0.99

Formula: C₁₂H₂₀O₂
Density: 0,99 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00135942
CAS Number: 5655-61-8

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR