Bindone ≥97.0% (by GC, titration analysis)
Synonyms:
[1,2'-Biindenylidene]-1',3,3'(2H)-trione
B0462-1G
TCB0462-001GEA
37.74
USD
TCB0462-001G
Bindone ≥97.0% (by GC, titration analysis)
Bindone
[for Detection of Primary Amines]
CAS Number: 1707-95-5
MDL Number: MFCD00021233
Molecular Formula: C18H10O3
Molecular Weight: 274.28
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 208
Other Product Names / Synonyms: [1,2'-Biindenylidene]-1',3,3'(2H)-trione, 2-(3-Oxo-1-indanylidene)indan-1,3-dione
Formula:
C₁₈H₁₀O₃ MW: 274.2702 g/mol Melting Pt: 208 °C |
MDL Number:
MFCD00021233 CAS Number: 1707-95-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...