Beryllon II
Supplier: TCI America
Synonyms:
2-(8-Hydroxy-3,6-disulfo-1-naphthylazo)-1,8-dihydroxynaphthalene-3,6-disulfonic acid tetrasodium salt
, 8-(1,8-Dihydroxy-3,6-disulpho-2-naphthylazo)-1-naphthol-3,6-disulphonic acid tetrasodium salt
, 2-(8-Hydroxy-3,6-disulpho-1-naphthylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid tetrasodium salt
, 8-(1,8-Dihydroxy-3,6-disulfo-2-naphthylazo)-1-naphthol-3,6-disulfonic acid tetrasodium salt
CAS Number: 51550-25-5
MDL Number: MFCD00046409
Molecular Formula: C20H14N2O15S4
Molecular Weight: 738.50
Form: Crystal
Color: Purple
Other Product Names / Synonyms: 8-(1,8-Dihydroxy-3,6-disulfo-2-naphthylazo)-1-naphthol-3,6-disulfonic Acid Tetrasodium Salt, 2-(8-Hydroxy-3,6-disulfo-1-naphthylazo)-1,8-dihydroxynaphthalene-3,6-disulfonic Acid Tetrasodium Salt
Formula:
C₂₀H₁₄N₂O₁₅S₄ MW: 738.50 g/mol |
MDL Number:
MFCD00046409 CAS Number: 51550-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...