1-Benzyl-1-phenylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Benzyl-N-phenylhydrazine hydrochloride

B0434-500G B0434-25G
TCB0434-500GEA 791.08 USD
TCB0434-500G TCB0434-025G
1-Benzyl-1-phenylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)
1-Benzyl-1-phenylhydrazine hydrochloride

CAS Number: 5705-15-7
MDL Number: MFCD00050690
Molecular Formula: C13H14N2
Molecular Weight: 234.73
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 176

Formula: C₁₃H₁₅ClN₂
Melting Pt: 176 °C
MDL Number: MFCD00050690
CAS Number: 5705-15-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR