Benzoylurea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Benzoylcarbamide

B0400-500G B0400-25G
TCB0400-500GEA 752 USD InStock
TCB0400-500G TCB0400-025G
Benzoylurea ≥98.0% (by HPLC, total nitrogen)
Benzoylurea

CAS Number: 614-22-2
MDL Number: MFCD00025441
Molecular Formula: C8H8N2O2
Molecular Weight: 164.16
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 210


Other Product Names / Synonyms: Benzoylcarbamide

Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00025441
CAS Number: 614-22-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR