Benzoyl-DL-alanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0135-25G
TCB0135-025GEA 53.87 USD InStock
TCB0135-025G
Benzoyl-DL-alanine ≥98.0% (by titrimetric analysis)
Benzoyl-DL-alanine

CAS Number: 1205-02-3
MDL Number: MFCD00020393
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 166

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00020393
CAS Number: 1205-02-3

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR