2'-Bromo-4'-methylacetanilide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

B0125-10G
TCB0125-010GEA 913.91 USD
TCB0125-010G
2'-Bromo-4'-methylacetanilide ≥98.0% (by total nitrogen basis)
2'-Bromo-4'-methylacetanilide

CAS Number: 614-83-5
MDL Number: MFCD00017781
Molecular Formula: C9H10BrNO
Molecular Weight: 228.09
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 119
Flash Point (°C): 113


Other Product Names / Synonyms: N-Acetyl-2-bromo-p-toluidine

https://us.vwr.com/stibo/web/std.lang.all/22/78/28932278.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR