Benzamidoxime ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Benzamide oxime
, N'-Hydroxybenzimidamide
B0015-25G
TCB0015-025GEA
197.93
USD
TCB0015-025G
Benzamidoxime ≥98.0% (by HPLC, titration analysis)
Benzamidoxime
CAS Number: 613-92-3
MDL Number: MFCD00031485
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 77
Formula:
C₇H₈N₂O MW: 136.15 g/mol Melting Pt: 77 °C |
MDL Number:
MFCD00031485 CAS Number: 613-92-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...