1-(3-Aminopropyl)pyrrolidine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-(Pyrrolidin-1-yl)propan-1-amine
, N-(3-Aminopropyl)pyrrolidine
A2270-25G
A2270-5G
TCA2270-5GEA
58.87
USD
TCA2270-5G
TCA2270-25G
1-(3-Aminopropyl)pyrrolidine ≥98.0% (by titrimetric analysis)
1-(3-Aminopropyl)pyrrolidine
CAS Number: 23159-07-1
MDL Number: MFCD00014100
Molecular Formula: C7H16N2
Molecular Weight: 128.22
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 82
Specific Gravity (20/20): 0.90
Formula:
C₇H₁₆N₂ MW: 128.22 g/mol Boiling Pt: 82 °C Density: 0.90 (20 °C) |
MDL Number:
MFCD00014100 CAS Number: 23159-07-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...