1-Adamantanecarboxamide ≥97.0%

Supplier: TCI America

Synonyms: Adamantane-1-carboxamide

A2078-5G A2078-1G
TCA2078-5GEA 136.99 USD
TCA2078-5G TCA2078-1G
1-Adamantanecarboxamide ≥97.0%
1-Adamantanecarboxamide

CAS Number: 5511-18-2
MDL Number: MFCD00077200
Molecular Formula: C11H17NO
Molecular Weight: 179.26
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 188

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00077200
CAS Number: 5511-18-2
https://us.vwr.com/stibo/web/std.lang.all/02/23/28930223.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR