6-Aminoindole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2067-5G A2067-1G
TCA2067-5GEA 210 USD InStock
TCA2067-5G TCA2067-1G
6-Aminoindole ≥98.0% (by GC, titration analysis)
6-Aminoindole

CAS Number: 5318-27-4
MDL Number: MFCD02093959
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 72
Storage Temperature: 0-10°C

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 166 °C
Melting Pt: 72 °C
Storage Temperature: -20°C
MDL Number: MFCD02093959
CAS Number: 5318-27-4
EINECS: 000-000-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR