Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-nitropyrimidine ≥98.0%

Supplier: TCI America


A2031-1G
TCA2031-1GEA 88.44 USD InStock
TCA2031-1G
2-Amino-5-nitropyrimidine ≥98.0%
2-Amino-5-nitropyrimidine
CAS Number: 3073-77-6
MDL Number: MFCD00006103
Molecular Formula: C4H4N4O2
Molecular Weight: 140.10
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 236


Formula: C₄H₄N₄O₂
MW: 140.1 g/mol
Melting Pt: 236 °C
Density: 1.558 g/cm³ (20 °C)
MDL Number: MFCD00006103
CAS Number: 3073-77-6
EINECS: 221-348-1
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty