4'-Acetoxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: p-Acetylphenyl acetate4-Acetoxyacetophenone4-Acetylphenyl acetate

A1997-25G
TCA1997-25GEA 34.31 USD
TCA1997-25G
4'-Acetoxyacetophenone ≥98.0%
4'-Acetoxyacetophenone

CAS Number: 13031-43-1
MDL Number: MFCD00017229
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 54

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 170 °C
Melting Pt: 54 °C
MDL Number: MFCD00017229
CAS Number: 13031-43-1
https://us.vwr.com/stibo/web/std.lang.all/01/38/28930138.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR