Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Acetoxyacetophenone ≥98.0%

Supplier: TCI America


A1995-25G A1995-5G
TCA1995-25GEA 117.56 USD InStock
TCA1995-25G TCA1995-5G
2'-Acetoxyacetophenone ≥98.0%
2'-Acetoxyacetophenone
CAS Number: 7250-94-4
MDL Number: MFCD00017223
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 90


Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 118 °C
Melting Pt: 90 °C
MDL Number: MFCD00017223
CAS Number: 7250-94-4
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty