Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-fluorobenzothiazole ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A1993-5G
TCA1993-5G
2-Amino-6-fluorobenzothiazole ≥97.0% (by HPLC, titration analysis)
CAS Number: 348-40-3
MDL Number: MFCD00013336
Molecular Formula: C7H5FN2S
Molecular Weight: 168.19
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 183


Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 183 °C
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00013336
CAS Number: 348-40-3
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (HPLC,T)
Melting point (°C) 183


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty