4-Amino-3-chlorobenzonitrile ≥98.0%

Supplier: TCI America

A1962-25G A1962-5G
TCA1962-25GEA 265.74 USD
TCA1962-25G TCA1962-5G
4-Amino-3-chlorobenzonitrile ≥98.0%
4-Amino-3-chlorobenzonitrile

CAS Number: 21803-75-8
MDL Number: MFCD00052917
Molecular Formula: C7H5ClN2
Molecular Weight: 152.58
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 104


Other Product Names / Synonyms: 2-Chloro-4-cyanoaniline

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00052917
CAS Number: 21803-75-8
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/00/89/28930089.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR