3-Aminobenzophenone ≥98.0% (by titrimetric analysis)

Synonyms: m-Aminobenzophenone

A1899-5G
TCA1899-5GEA 88.74 USD
TCA1899-5G
3-Aminobenzophenone ≥98.0% (by titrimetric analysis)
3-Aminobenzophenone

CAS Number: 2835-78-1
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 86

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00017103
CAS Number: 2835-78-1
https://us.vwr.com/stibo/web/std.lang.all/99/92/28929992.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR