3-Aminophenylboronic acid hemisulfate ≥98%
Supplier: TCI America
Synonyms:
(3-Aminophenyl)boronic acid sulfate (2:1)
, 3-Aminobenzeneboronic acid hemisulphate
, 3-Aminobenzeneboronic acid hemisulfate
, (3-Aminophenyl)boronic acid sulphate (2:1)
A1774-1G
A1774-25G
A1774-5G
TCA1774-5GEA
95.41
USD
TCA1774-5G
TCA1774-1G
TCA1774-25G
3-Aminophenylboronic acid hemisulfate ≥98%
3-Aminophenylboronic acid hemisulphate
CAS Number: 66472-86-4
Molecular Formula: C6H8BNO2
Molecular Weight: 185.98
Form: Crystal
Color: White
Other Product Names / Synonyms: 3-Aminobenzeneboronic Acid Hemisulfate
Formula:
C₁₂H₁₈B₂N₂O₈S MW: 371,97 g/mol |
MDL Number:
MFCD00013111 CAS Number: 66472-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...