(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S,S)-TsDPEN
, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide
, N-((1S,2S)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulphonamide
, (1S,2S)-(+)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine
, (S,S)-N-(p-Toluenesulfonyl)-1,2-diphenylethylenediamine
, (1S,2S)-(+)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine
, (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethylenediamine
CAS Number: 167316-27-0
MDL Number: MFCD03095684
Molecular Formula: C21H22N2O2S
Molecular Weight: 366.48
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 130
Specific rotation [a]20/D: 30 deg (C=1, CH3CN)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-N-(p-Toluenesulfonyl)-1,2-diphenylethylenediamine
Formula:
C₂₁H₂₂N₂O₂S MW: 366.48 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD03095684 CAS Number: 167316-27-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...