Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminoanthracene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Anthrylamine

A1687-1G
TCA1687-1GEA 163.35 USD InStock
TCA1687-1G
2-Aminoanthracene ≥98.0% (by GC, titration analysis)
2-Aminoanthracene
CAS Number: 613-13-8
MDL Number: MFCD00003582
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal

Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2-Anthramine


Formula: C₁₄H₁₁N
MW: 193,25 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 218 to 219 °C
Density: Min. 1 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00003582
CAS Number: 613-13-8
EINECS: 210-330-9
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty