Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-4-(methylthio)benzotrifluoride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methylthio-5-(trifluoromethyl)aniline

A1666-1G
TCA1666-1GEA 108.2 USD InStock
TCA1666-1G
3-Amino-4-(methylthio)benzotrifluoride ≥98.0% (by GC, titration analysis)
3-Amino-4-(methylthio)benzotrifluoride
CAS Number: 207974-07-0
MDL Number: MFCD00052715
Molecular Formula: C8H8F3NS
Molecular Weight: 207.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 112
Specific Gravity (20/20): 1.34

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Amino-1-methylthio-4-trifluoromethylbenzene, 2-(Methylthio)-5-trifluoromethylaniline, alpha,alpha,alpha-Trifluoro-6-(methylthio)-m-toluidine


Formula: C₈H₈F₃NS
MW: 207,22
Boiling Pt: 112 °C
Density: 1.34 (20 °C)
MDL Number: MFCD00052715
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty