4-Amino-3-nitrobenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (4-Amino-3-nitrophenyl)(phenyl)methanone

A1644-25G
TCA1644-25GEA 87.26 USD
TCA1644-25G
4-Amino-3-nitrobenzophenone ≥98.0%
4-Amino-3-nitrobenzophenone

CAS Number: 31431-19-3
MDL Number: MFCD00007154
Molecular Formula: C13H10N2O3
Molecular Weight: 242.23
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 141

Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00007154
CAS Number: 31431-19-3
https://us.vwr.com/stibo/web/std.lang.all/98/09/28929809.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR