N-Acetyl-L-tyrosine ≥99.0% (by titrimetric analysis)
Synonyms:
Ac-Tyr-OH
A1409-25G
TCA1409-025GEA
41.2
USD
TCA1409-025G
N-Acetyl-L-tyrosine ≥99.0% (by titrimetric analysis)
N-Acetyl-L-tyrosine
CAS Number: 537-55-3
MDL Number: MFCD00037190
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 152
Specific rotation [a]20/D: 47 deg (C=2, H2O)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Tyr-OH
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol Melting Pt: 152 °C |
MDL Number:
MFCD00037190 CAS Number: 537-55-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...