3'-Acetamidoacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
3-Acetamidoacetophenone
A1408-500G
A1408-25G
TCA1408-025GEA
67
USD
TCA1408-025G
TCA1408-500G
3'-Acetamidoacetophenone ≥98.0%
3'-Acetamidoacetophenone
CAS Number: 7463-31-2
MDL Number: MFCD00032278
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 127
Other Product Names / Synonyms: 3'-Acetylacetanilide
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...