Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Aminoethyl)benzenesulphonamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America


A1363-25G
TCA1363-025GEA 65.97 USD InStock
TCA1363-025G
4-(2-Aminoethyl)benzenesulphonamide ≥98.0% (by HPLC, titration analysis)
4-(2-Aminoethyl)benzenesulphonamide
CAS Number: 35303-76-5
MDL Number: MFCD00010301
Molecular Formula: C8H12N2O2S
Molecular Weight: 200.26
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 149


Formula: C₈H₁₂N₂O₂S
MW: 200,26
Melting Pt: 149 °C
MDL Number: MFCD00010301
CAS Number: 35303-76-5
EINECS: 252-501-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty