2'-Amino-4',5'-methylenedioxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-(6-Aminobenzo[d][1,3]dioxol-5-yl)ethanone
A1356-5G
TCA1356-5GEA
107.35
USD
TCA1356-5G
2'-Amino-4',5'-methylenedioxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Amino-4',5'-methylenedioxyacetophenone
CAS Number: 28657-75-2
MDL Number: MFCD00075855
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 171
Other Product Names / Synonyms: 5-Acetyl-6-amino-1,3-benzodioxole
Formula:
C₉H₉NO₃ MW: 179.1727 g/mol Melting Pt: 171 °C |
MDL Number:
MFCD00075855 CAS Number: 28657-75-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...