7-Aminocephalosporanic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
7-ACA
A1266-25G
A1266-5G
TCA1266-25GEA
139.79
USD
TCA1266-25G
TCA1266-005G
7-Aminocephalosporanic acid ≥97.0% (by HPLC, titration analysis)
7-Aminocephalosporanic acid
CAS Number: 957-68-6
MDL Number: MFCD00005177
Molecular Formula: C10H12N2O5S
Molecular Weight: 272.28
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 90 deg (C=0.5, buffer sol. pH7.0 KH2PO4)
Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: 7-ACA
Formula:
C₁₀H₁₂N₂O₅S MW: 272.28 g/mol |
MDL Number:
MFCD00005177 CAS Number: 957-68-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...