(S)-(-)-3-Acetamidopyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A1170-5G
TCA1170-005GEA
162
USD
TCA1170-005G
(S)-(-)-3-Acetamidopyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-3-Acetamidopyrrolidine
CAS Number: 114636-31-6
MDL Number: MFCD00082748
Molecular Formula: C6H12N2O
Molecular Weight: 128.18
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -48 deg (C=1, EtOH)
Storage Temperature: 0-10°C
Formula:
C₆H₁₂N₂O |
MDL Number:
MFCD00082748 CAS Number: 114636-31-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...