5-Acetylsalicylamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Acetyl-2-hydroxybenzamide

A1156-25G
TCA1156-025GEA 44.64 USD
TCA1156-025G
5-Acetylsalicylamide ≥98.0% (by titrimetric analysis)
5-Acetylsalicylamide

CAS Number: 40187-51-7
MDL Number: MFCD00049222
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 224


Other Product Names / Synonyms: 5-Acetyl-2-hydroxybenzamide

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 224 °C
MDL Number: MFCD00049222
CAS Number: 40187-51-7
https://us.vwr.com/stibo/web/std.lang.all/90/01/28929001.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR